Product Details

SNP ID
rs12738679
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:59827211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAATTTGCTCTTTCTTTGGGGG[A/G]GTTCAGCCTTTTTCTTAAGGCCTTC
Phenotype
MIM: 607820
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOOK1 PubMed Links

Gene Details

Gene
HOOK1
Gene Name
hook microtubule-tethering protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015888.4 Intron NP_056972.1
XM_006710676.1 Intron XP_006710739.1
XM_011541562.2 Intron XP_011539864.1
XM_011541563.1 Intron XP_011539865.1
XM_017001424.1 Intron XP_016856913.1

View Full Product Details