Product Details

SNP ID
rs6715676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:26411344 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATATAAAAGTATTATTTTAATAT[A/G]CAATCAGTGTAAATCATACTGATGA
Phenotype
MIM: 615288
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DRC1 PubMed Links

Gene Details

Gene
DRC1
Gene Name
dynein regulatory complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145038.4 Intron NP_659475.2
XM_005264637.3 Intron XP_005264694.1
XM_005264638.3 Intron XP_005264695.1
XM_017005271.1 Intron XP_016860760.1

View Full Product Details