Product Details
- SNP ID
-
hCV30786580
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:200878757 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTCCTTCTGAATATAATTACAT[C/G]GTAACATTTTAGTCTTCTGGGAAAT
- Phenotype
-
MIM: 615811
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PPIL3
PubMed Links
Gene Details
- Gene
- PPIL3
- Gene Name
- peptidylprolyl isomerase like 3
There are no transcripts associated with this gene.
View Full Product Details