Product Details
- SNP ID
-
rs11685853
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:200817672 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTTTTAAAAAACTGCTTGCAGTA[A/G]TGAGTATTTAAGTTACCTTACTGTA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BZW1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs191066589] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BZW1
- Gene Name
- basic leucine zipper and W2 domains 1
- Gene
- LOC101927795
- Gene Name
- uncharacterized LOC101927795
There are no transcripts associated with this gene.
View Full Product Details