Product Details

SNP ID
rs11105887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:90964578 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTGAGAAAACTACTGAAAGAAAA[A/G]CAAGTTATTTCCAAAAATGATGAGT
Phenotype
MIM: 601657
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCER1 PubMed Links

Gene Details

Gene
CCER1
Gene Name
coiled-coil glutamate rich protein 1
There are no transcripts associated with this gene.

Gene
EPYC
Gene Name
epiphycan
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004950.4 Intron NP_004941.2
XM_011538008.1 Intron XP_011536310.1

View Full Product Details