Product Details

SNP ID
rs10752254
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:12080356 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGTGCATAGAATACAAACCCAAG[C/T]GCATGGGTGGCTCTGAAATCATTCC
Phenotype
MIM: 614984
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHTKD1 PubMed Links

Gene Details

Gene
DHTKD1
Gene Name
dehydrogenase E1 and transketolase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018706.6 Intron NP_061176.3

View Full Product Details