Product Details

SNP ID
rs12708540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:36583321 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTAGTAAATGCTGAGAGGTGAA[A/G]ATGTAGCTTAATTTTTTCTCCAGCT
Phenotype
MIM: 615626
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C15orf41 PubMed Links

Gene Details

Gene
C15orf41
Gene Name
chromosome 15 open reading frame 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130010.2 Intron NP_001123482.1
NM_001290232.1 Intron NP_001277161.1
NM_001290233.1 Intron NP_001277162.1
NM_001321756.1 Intron NP_001308685.1
NM_001321757.1 Intron NP_001308686.1
NM_001321758.1 Intron NP_001308687.1
NM_001321759.1 Intron NP_001308688.1
NM_001321760.1 Intron NP_001308689.1
NM_001321761.1 Intron NP_001308690.1
NM_032499.5 Intron NP_115888.1
XM_011522111.1 Intron XP_011520413.1
XM_017022676.1 Intron XP_016878165.1
XM_017022677.1 Intron XP_016878166.1

View Full Product Details