Product Details

SNP ID
rs10779902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:99310151 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAATGCCTGGCATGTAAATGTTA[C/G]TTGTTAACTAAAAATAAATAGGTCT
Phenotype
MIM: 612564
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LYG1 PubMed Links

Gene Details

Gene
LYG1
Gene Name
lysozyme g1
There are no transcripts associated with this gene.

Gene
TXNDC9
Gene Name
thioredoxin domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005783.3 Intron NP_005774.2
XM_017003147.1 Intron XP_016858636.1

View Full Product Details