Product Details

SNP ID
rs7831911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:70642481 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCTTTTTCCTTTTGGGCCTTTTT[A/G]TTGGGTCCTGTGTCTTTCTTAGCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LACTB2 PubMed Links

Gene Details

Gene
LACTB2
Gene Name
lactamase beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016027.2 Intron NP_057111.1
Gene
LACTB2-AS1
Gene Name
LACTB2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details