Product Details

SNP ID
rs12352904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120392164 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGAGCATCTCCTCTGCCCCCGAGA[C/T]TGTGCTAAGTACAATACAGTTTAGG
Phenotype
MIM: 608201
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK5RAP2 PubMed Links

Gene Details

Gene
CDK5RAP2
Gene Name
CDK5 regulatory subunit associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011649.2 Intron NP_001011649.1
NM_001272039.1 Intron NP_001258968.1
NM_018249.5 Intron NP_060719.4
XM_006717182.1 Intron XP_006717245.1
XM_006717185.1 Intron XP_006717248.1
XM_011518860.1 Intron XP_011517162.1
XM_017014921.1 Intron XP_016870410.1
XM_017014922.1 Intron XP_016870411.1
XM_017014923.1 Intron XP_016870412.1
XM_017014924.1 Intron XP_016870413.1

View Full Product Details