Product Details

SNP ID
rs13186619
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157480125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTCACCAAAGCACCACACATTA[A/G]AGGGAGAATGAGAGGGGAAGGAAGA
Phenotype
MIM: 603640 MIM: 609383
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM19 PubMed Links

Gene Details

Gene
ADAM19
Gene Name
ADAM metallopeptidase domain 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033274.4 2925 UTR 3 NP_150377.1
XM_005266003.2 2925 Intron XP_005266060.1
XM_011534682.2 2925 UTR 3 XP_011532984.1
XM_017010009.1 2925 UTR 3 XP_016865498.1
Gene
NIPAL4
Gene Name
NIPA like domain containing 4
There are no transcripts associated with this gene.

View Full Product Details