Product Details

SNP ID
rs11618185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:51863344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTGTGTGAACTGATGTGATAGG[A/C]CGAGAGAGTGTTCTAGAAAACACCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC70 PubMed Links

Gene Details

Gene
CCDC70
Gene Name
coiled-coil domain containing 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031290.2 Intron NP_112580.2

View Full Product Details