Product Details

SNP ID
rs13423806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218785573 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTAGAGCAGTAACTAACCAAAGA[A/C]ATTCTAGATGGCTTTAGGATGAGGA
Phenotype
MIM: 606530
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CYP27A1 PubMed Links

Gene Details

Gene
CYP27A1
Gene Name
cytochrome P450 family 27 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000784.3 Intron NP_000775.1
XM_017003488.1 Intron XP_016858977.1

View Full Product Details