Product Details

SNP ID
rs12637949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:136873345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGATTTGCTAGAGGCCTTTAAC[A/C]CCTTTGTTCTGGGCAATTTCTCCCA
Phenotype
MIM: 600508
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NCK1 PubMed Links

Gene Details

Gene
NCK1
Gene Name
NCK adaptor protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190796.2 Intron NP_001177725.1
NM_001291999.1 Intron NP_001278928.1
NM_006153.5 Intron NP_006144.1

View Full Product Details