Product Details

SNP ID
rs11580421
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:40696884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATGCATGCACTGTCACAGTTTA[A/G]GAAGGCTACAAGGTCATTTTAGTGT
Phenotype
MIM: 605344 MIM: 611600
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NFYC PubMed Links

Gene Details

Gene
NFYC
Gene Name
nuclear transcription factor Y subunit gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142587.1 Intron NP_001136059.1
NM_001142588.1 Intron NP_001136060.1
NM_001142589.1 Intron NP_001136061.1
NM_001142590.1 Intron NP_001136062.1
NM_001308114.1 Intron NP_001295043.1
NM_001308115.1 Intron NP_001295044.1
NM_014223.4 Intron NP_055038.2
XM_005270894.1 Intron XP_005270951.1
XM_005270895.3 Intron XP_005270952.3
XM_006710658.2 Intron XP_006710721.2
XM_006710660.2 Intron XP_006710723.2
XM_006710661.1 Intron XP_006710724.1
XM_006710662.2 Intron XP_006710725.2
XM_011541516.1 Intron XP_011539818.1
XM_011541517.1 Intron XP_011539819.1
XM_017001364.1 Intron XP_016856853.1
XM_017001365.1 Intron XP_016856854.1
XM_017001366.1 Intron XP_016856855.1
XM_017001367.1 Intron XP_016856856.1
XM_017001368.1 Intron XP_016856857.1
XM_017001369.1 Intron XP_016856858.1
Gene
NFYC-AS1
Gene Name
NFYC antisense RNA 1
There are no transcripts associated with this gene.

Gene
RIMS3
Gene Name
regulating synaptic membrane exocytosis 3
There are no transcripts associated with this gene.

View Full Product Details