Product Details

SNP ID
rs12515331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:170380045 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGCAAACAAACTATGTTGGCAAG[C/T]GGGTATTTGACACTGGGATCCAAAG
Phenotype
MIM: 604660 MIM: 603951
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTD-2270F17.1 PubMed Links

Gene Details

Gene
CTD-2270F17.1
Gene Name
uncharacterized LOC101928033
There are no transcripts associated with this gene.

Gene
KCNIP1
Gene Name
potassium voltage-gated channel interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034837.2 Intron NP_001030009.1
NM_001034838.2 Intron NP_001030010.1
NM_001278339.1 Intron NP_001265268.1
NM_001278340.1 Intron NP_001265269.1
NM_014592.3 Intron NP_055407.1
XM_017009407.1 Intron XP_016864896.1
XM_017009408.1 Intron XP_016864897.1
Gene
KCNMB1
Gene Name
potassium calcium-activated channel subfamily M regulatory beta subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004137.3 Intron NP_004128.1

View Full Product Details