Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001102609.2 | 761 | Intron | NP_001096079.1 | ||
NM_001305393.1 | 761 | Intron | NP_001292322.1 | ||
NM_001305394.1 | 761 | Intron | NP_001292323.1 | ||
NM_001305395.1 | 761 | Intron | NP_001292324.1 | ||
NM_001305396.1 | 761 | Intron | NP_001292325.1 | ||
NM_001305397.1 | 761 | Intron | NP_001292326.1 | ||
XM_011534432.2 | 761 | UTR 5 | XP_011532734.1 | ||
XM_017009029.1 | 761 | UTR 5 | XP_016864518.1 | ||
XM_017009030.1 | 761 | UTR 5 | XP_016864519.1 | ||
XM_017009031.1 | 761 | Intron | XP_016864520.1 | ||
XM_017009032.1 | 761 | Intron | XP_016864521.1 | ||
XM_017009033.1 | 761 | Intron | XP_016864522.1 |