Product Details

SNP ID
rs12266417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:72897530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATATATACCAAAATTCACTTTGA[A/G]AATATTTCTGGTAACAACATGCTAG
Phenotype
MIM: 614197 MIM: 609330
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MCU PubMed Links

Gene Details

Gene
MCU
Gene Name
mitochondrial calcium uniporter
There are no transcripts associated with this gene.

Gene
OIT3
Gene Name
oncoprotein induced transcript 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152635.2 Intron NP_689848.1

View Full Product Details