Product Details

SNP ID
rs11570117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47331972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCAGGGTCCGTGCCCTTGCAGC[C/T]AGGGCAGGTGCTTGGCCGAGGACAA
Phenotype
MIM: 603584 MIM: 600958
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MADD PubMed Links

Gene Details

Gene
MADD
Gene Name
MAP kinase activating death domain
There are no transcripts associated with this gene.

Gene
MYBPC3
Gene Name
myosin binding protein C, cardiac
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000256.3 Intron NP_000247.2

View Full Product Details