Product Details

SNP ID
rs12682180
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38263465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACAGTTATGGTCAAATGAGGTA[A/G]AAACAGTCATCTGTTAGTAGTGCTG
Phenotype
MIM: 615003 MIM: 610626
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DDHD2 PubMed Links

Gene Details

Gene
DDHD2
Gene Name
DDHD domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164232.1 2135 Intron NP_001157704.1
NM_001164234.1 2135 Intron NP_001157706.1
NM_015214.2 2135 Intron NP_056029.2
XM_005273454.2 2135 Intron XP_005273511.1
XM_005273455.3 2135 Intron XP_005273512.1
XM_005273456.3 2135 Intron XP_005273513.1
XM_011544456.1 2135 Intron XP_011542758.1
XM_017013252.1 2135 Intron XP_016868741.1
XM_017013253.1 2135 Intron XP_016868742.1
XM_017013254.1 2135 Intron XP_016868743.1
XM_017013255.1 2135 Intron XP_016868744.1
XM_017013256.1 2135 Intron XP_016868745.1
Gene
PLPP5
Gene Name
phospholipid phosphatase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102559.1 2135 UTR 3 NP_001096029.1
NM_001102560.1 2135 Intron NP_001096030.1
NM_032483.3 2135 Intron NP_115872.2
XM_005273661.4 2135 Intron XP_005273718.2
XM_005273663.3 2135 Intron XP_005273720.2
XM_011544670.2 2135 Intron XP_011542972.1
XM_011544671.1 2135 UTR 3 XP_011542973.1
XM_011544672.1 2135 UTR 3 XP_011542974.1
XM_011544674.2 2135 Intron XP_011542976.1
XM_011544675.2 2135 Intron XP_011542977.1
XM_017013905.1 2135 Intron XP_016869394.1
XM_017013906.1 2135 Intron XP_016869395.1
XM_017013907.1 2135 Intron XP_016869396.1

View Full Product Details