Product Details

SNP ID
rs10832948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18607183 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGCCAGACTTACTTTTTTGTTAAC[A/G]GTCTTTGAACCTTTTCACGGCACAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPTY2D1 PubMed Links

Gene Details

Gene
SPTY2D1
Gene Name
SPT2 chromatin protein domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194285.2 4810 UTR 3 NP_919261.2
XM_011519919.2 4810 UTR 3 XP_011518221.1
Gene
SPTY2D1-AS1
Gene Name
SPTY2D1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details