Product Details

SNP ID
rs11098431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:118281172 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAGGTGCTTGTTAAAAGCAAATA[C/T]AGTAGATATGTTAATGTGTTGGATA
Phenotype
MIM: 606709
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRSS12 PubMed Links

Gene Details

Gene
PRSS12
Gene Name
protease, serine 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003619.3 3439 UTR 3 NP_003610.2
XM_005263318.4 3439 Intron XP_005263375.1
XM_011532387.2 3439 Intron XP_011530689.1
Gene
SNHG8
Gene Name
small nucleolar RNA host gene 8
There are no transcripts associated with this gene.

Gene
SNORA24
Gene Name
small nucleolar RNA, H/ACA box 24
There are no transcripts associated with this gene.

View Full Product Details