Product Details

SNP ID
rs12440537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89472602 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGGCAGGAGGGGTGGAAGCCCTC[A/C]TTTCTTGATGTTTGCTTTTTGCCTC
Phenotype
MIM: 605381
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC105371031 PubMed Links

Gene Details

Gene
LOC105371031
Gene Name
translation initiation factor IF-2-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017022800.1 Intron XP_016878289.1
Gene
RHCG
Gene Name
Rh family C glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321041.1 Intron NP_001307970.1
NM_016321.2 Intron NP_057405.1

View Full Product Details