Product Details

SNP ID
rs8068517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18192664 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTCAACTCACTAACTCCTGTGTG[A/G]TACTTCTCACTGTGGGCTGATGGGT
Phenotype
MIM: 613303
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALKBH5 PubMed Links

Gene Details

Gene
ALKBH5
Gene Name
alkB homolog 5, RNA demethylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017758.3 Intron NP_060228.3

View Full Product Details