Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001318736.1 | 5826 | Intron | NP_001305665.1 | ||
NM_001318737.1 | 5826 | Intron | NP_001305666.1 | ||
NM_024725.3 | 5826 | Intron | NP_079001.2 | ||
XM_005274292.3 | 5826 | Intron | XP_005274349.1 | ||
XM_006718911.3 | 5826 | Intron | XP_006718974.1 | ||
XM_011542984.2 | 5826 | Intron | XP_011541286.1 | ||
XM_011542987.2 | 5826 | Intron | XP_011541289.1 | ||
XM_011542988.2 | 5826 | Intron | XP_011541290.1 | ||
XM_017018305.1 | 5826 | Intron | XP_016873794.1 | ||
XM_017018306.1 | 5826 | Intron | XP_016873795.1 | ||
XM_017018307.1 | 5826 | Intron | XP_016873796.1 | ||
XM_017018308.1 | 5826 | Intron | XP_016873797.1 | ||
XM_017018309.1 | 5826 | UTR 3 | XP_016873798.1 | ||
XM_017018310.1 | 5826 | UTR 3 | XP_016873799.1 | ||
XM_017018311.1 | 5826 | Intron | XP_016873800.1 | ||
XM_017018312.1 | 5826 | Intron | XP_016873801.1 |