Product Details

SNP ID
rs13407714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241074405 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGATTCTCTTAATTCAGGTGTGG[C/T]CCATTCAGAGTGAAGACTCTCGTGT
Phenotype
MIM: 615393 MIM: 616634
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTERF4 PubMed Links

Gene Details

Gene
MTERF4
Gene Name
mitochondrial transcription termination factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182501.3 4517 Intron NP_872307.2
XM_006712292.3 4517 Intron XP_006712355.1
XM_006712294.1 4517 Intron XP_006712357.1
XM_006712295.3 4517 Intron XP_006712358.1
XM_011510634.2 4517 Intron XP_011508936.1
XM_017003379.1 4517 Intron XP_016858868.1
XM_017003380.1 4517 Intron XP_016858869.1
XM_017003381.1 4517 Intron XP_016858870.1
XM_017003382.1 4517 Intron XP_016858871.1
XM_017003383.1 4517 Intron XP_016858872.1
XM_017003384.1 4517 Intron XP_016858873.1
XM_017003385.1 4517 UTR 3 XP_016858874.1
XM_017003386.1 4517 Intron XP_016858875.1
XM_017003387.1 4517 Intron XP_016858876.1
XM_017003388.1 4517 Intron XP_016858877.1
XM_017003389.1 4517 Intron XP_016858878.1
XM_017003390.1 4517 Intron XP_016858879.1
XM_017003391.1 4517 Intron XP_016858880.1
Gene
SNED1
Gene Name
sushi, nidogen and EGF like domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080437.1 4517 Intron NP_001073906.1
XM_011510931.2 4517 Intron XP_011509233.1
XM_011510932.2 4517 Intron XP_011509234.1
XM_011510933.2 4517 Intron XP_011509235.1
XM_011510934.2 4517 Intron XP_011509236.1
XM_011510937.2 4517 Intron XP_011509239.1
XM_017003773.1 4517 Intron XP_016859262.1
XM_017003774.1 4517 Intron XP_016859263.1
XM_017003775.1 4517 Intron XP_016859264.1
XM_017003776.1 4517 Intron XP_016859265.1
XM_017003777.1 4517 Intron XP_016859266.1
XM_017003778.1 4517 Intron XP_016859267.1
XM_017003779.1 4517 Intron XP_016859268.1
XM_017003780.1 4517 Intron XP_016859269.1
XM_017003781.1 4517 Intron XP_016859270.1
XM_017003782.1 4517 Intron XP_016859271.1

View Full Product Details