Product Details

SNP ID
rs7014211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42551660 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTAAACATTAATTTTAATAGCTTA[A/G]GTCAGGCATGATGGCTCACACCTGT
Phenotype
MIM: 158378
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC20A2 PubMed Links

Gene Details

Gene
SLC20A2
Gene Name
solute carrier family 20 member 2
There are no transcripts associated with this gene.

Gene
SMIM19
Gene Name
small integral membrane protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135674.1 Intron NP_001129146.1
NM_001135675.1 Intron NP_001129147.1
NM_001135676.1 Intron NP_001129148.1
NM_138436.3 Intron NP_612445.2
XM_005273398.4 Intron XP_005273455.1

View Full Product Details