Product Details

SNP ID
hCV30948679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22031342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTACCTTGGGCCTCTCACTAAG[A/G]GTATAGCCCTTGAGGGTCCTACCCT
Phenotype
MIM: 603725 MIM: 608073
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGF17 PubMed Links

Gene Details

Gene
FGF17
Gene Name
fibroblast growth factor 17
There are no transcripts associated with this gene.

Gene
NPM2
Gene Name
nucleophosmin/nucleoplasmin 2
There are no transcripts associated with this gene.

View Full Product Details