Product Details

SNP ID
rs12366563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56772166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTTTATTCTCTCTAGGAAGCTT[C/T]TCAGACCTCTCTAAACTGCCTTAGA
Phenotype
MIM: 606623
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSD17B6 PubMed Links

Gene Details

Gene
HSD17B6
Gene Name
hydroxysteroid 17-beta dehydrogenase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003725.3 Intron NP_003716.2
XM_005269207.1 Intron XP_005269264.1
XM_005269208.1 Intron XP_005269265.1
XM_005269209.1 Intron XP_005269266.1
XM_006719672.1 Intron XP_006719735.1
XM_011538925.1 Intron XP_011537227.1
XM_011538926.1 Intron XP_011537228.1
XM_011538927.1 Intron XP_011537229.1

View Full Product Details