Product Details

SNP ID
rs11057572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:124298275 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGACGTGCATGTTAAACCTGCCTA[A/C]GCTTCACTTCATTAAAGAGGCCCCC
Phenotype
MIM: 615927
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM101A PubMed Links

Gene Details

Gene
FAM101A
Gene Name
family with sequence similarity 101 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181709.4 Intron NP_859060.3
Gene
LOC105370045
Gene Name
filamin-interacting protein FAM101A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011539088.1 Intron XP_011537390.1
Gene
ZNF664-FAM101A
Gene Name
filamin-interacting protein FAM101A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204299.1 Intron NP_001191228.1

View Full Product Details