Product Details

SNP ID
rs13241841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:124824915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAATTGGATAATTGGAGTTTGTT[A/C]CAGAAAATGTTTACTGCCAACAATT
Phenotype
MIM: 606478
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
POT1 PubMed Links

Gene Details

Gene
POT1
Gene Name
protection of telomeres 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042594.1 Intron NP_001036059.1
NM_015450.2 Intron NP_056265.2
XM_006715917.3 Intron XP_006715980.1
XM_011516006.1 Intron XP_011514308.1
XM_011516007.1 Intron XP_011514309.1
XM_017011941.1 Intron XP_016867430.1
XM_017011942.1 Intron XP_016867431.1

View Full Product Details