Product Details

SNP ID
rs12129753
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:240489914 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGAATTATTTCCTAGGAAGTTGA[A/G]TTTAATTCCTTTGAAGCCTACACAG
Phenotype
MIM: 608832
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GREM2 PubMed Links

Gene Details

Gene
GREM2
Gene Name
gremlin 2, DAN family BMP antagonist
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022469.3 3842 UTR 3 NP_071914.3
XM_005273226.3 3842 UTR 3 XP_005273283.1
XM_011544249.2 3842 UTR 3 XP_011542551.1

View Full Product Details