Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001319677.1 | Intron | NP_001306606.1 | |||
NM_001319678.1 | Intron | NP_001306607.1 | |||
NM_001319679.1 | Intron | NP_001306608.1 | |||
NM_018056.3 | Intron | NP_060526.2 | |||
XM_005270988.2 | Intron | XP_005271045.1 | |||
XM_006710724.1 | Intron | XP_006710787.1 | |||
XM_006710725.3 | Intron | XP_006710788.1 | |||
XM_011541681.2 | Intron | XP_011539983.1 | |||
XM_011541683.1 | Intron | XP_011539985.1 | |||
XM_011541684.2 | Intron | XP_011539986.1 | |||
XM_017001595.1 | Intron | XP_016857084.1 |