Product Details

SNP ID
rs11887633
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:110114675 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTAACCCGTTCGCCAGTTTGCCTA[A/T]AAGTGGCTTGAGGGGGCCCAACTTT
Phenotype
MIM: 602022 MIM: 607100
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
MALL PubMed Links

Gene Details

Gene
MALL
Gene Name
mal, T-cell differentiation protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005434.4 Intron NP_005425.1
XM_011511809.1 Intron XP_011510111.1
Gene
NPHP1
Gene Name
nephrocystin 1
There are no transcripts associated with this gene.

View Full Product Details