Product Details

SNP ID
rs13388539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:110095680 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTTTAACCAATAAATCTAATCG[C/T]GTCATCATTTAAAAAAAGAAAAAAA
Phenotype
MIM: 602022
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MALL PubMed Links

Gene Details

Gene
MALL
Gene Name
mal, T-cell differentiation protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005434.4 Intron NP_005425.1
XM_011511809.1 Intron XP_011510111.1
Gene
MIR4436B1
Gene Name
microRNA 4436b-1
There are no transcripts associated with this gene.

View Full Product Details