Product Details

SNP ID
rs8056811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70357763 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTATATACATGCCACTGTGCTGG[G/A]TGTTACAAGGAACACAAGACCTACT
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
DDX19A PubMed Links

Gene Details

Gene
DDX19A
Gene Name
DEAD-box helicase 19A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320522.1 Intron NP_001307451.1
NM_001320525.1 Intron NP_001307454.1
NM_001320526.1 Intron NP_001307455.1
NM_001320527.1 Intron NP_001307456.1
NM_018332.4 Intron NP_060802.1

View Full Product Details