Product Details

SNP ID
rs12608893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49773157 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGGGCAAATTCTGCAGATAAGGA[C/G]GGGGAGAGGTTGAGGCTGGGCCCAG
Phenotype
MIM: 601026 MIM: 608253
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AP2A1 PubMed Links

Gene Details

Gene
AP2A1
Gene Name
adaptor related protein complex 2 alpha 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014203.2 Intron NP_055018.2
NM_130787.2 Intron NP_570603.2
XM_011526556.2 Intron XP_011524858.1
XM_011526557.2 Intron XP_011524859.1
Gene
TSKS
Gene Name
testis specific serine kinase substrate
There are no transcripts associated with this gene.

View Full Product Details