Product Details

SNP ID
rs12088105
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:154483107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAATTTTTGTTGTACCTTTTATT[C/T]TATAATTCAGAATTCTAATAATGAT
Phenotype
MIM: 610482
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SHE PubMed Links

Gene Details

Gene
SHE
Gene Name
Src homology 2 domain containing E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010846.2 2554 UTR 3 NP_001010846.1
XM_005244891.4 2554 Intron XP_005244948.1
XM_011509163.2 2554 Intron XP_011507465.1
XM_017000256.1 2554 Intron XP_016855745.1

View Full Product Details