Product Details

SNP ID
rs13233880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143913200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTCATACTCATTTCATTATCAG[G/T]GAAGTTGACCTTGTTTTCATGCATT
Phenotype
MIM: 608497
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR2F1 PubMed Links

Gene Details

Gene
OR2F1
Gene Name
olfactory receptor family 2 subfamily F member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012369.2 Intron NP_036501.2
XM_011516019.2 Intron XP_011514321.1
XM_011516020.2 Intron XP_011514322.1

View Full Product Details