Product Details

SNP ID
rs12037380
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:108857424 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGAATCTGATGGACTCAATTAAG[A/C]ATTTCTACAGATGGGAAAACCAAAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AKNAD1 PubMed Links

Gene Details

Gene
AKNAD1
Gene Name
AKNA domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152763.4 Intron NP_689976.2
Gene
SPATA42
Gene Name
spermatogenesis associated 42 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details