Product Details
- SNP ID
-
rs11490903
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:108972590 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTCAGGAGTATATGGGAAATCTC[C/T]GTATCTCTCAATTTTGCTGTTAACC
- Phenotype
-
MIM: 615734
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CLCC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77006276] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLCC1
- Gene Name
- chloride channel CLIC like 1
There are no transcripts associated with this gene.
- Gene
- WDR47
- Gene Name
- WD repeat domain 47
View Full Product Details