Product Details

SNP ID
rs12335678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97279419 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATATGAGTGGGATCCCTGCTTTCA[A/G]TTTCCAAGCAGAATGATTGGAACCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100499484 PubMed Links

Gene Details

Gene
LOC100499484
Gene Name
SUGT1-1300002K09Rik pseudogene
There are no transcripts associated with this gene.

Gene
LOC100499484-C9ORF174
Gene Name
LOC100499484-C9orf174 readthrough
There are no transcripts associated with this gene.

View Full Product Details