Product Details

SNP ID
rs11704764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23742685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTGTCCTCAGTCACCATGACTG[A/C]TCACAGTTGCCAAAGCACTTTCACA
Phenotype
MIM: 194544
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF70 PubMed Links

Gene Details

Gene
ZNF70
Gene Name
zinc finger protein 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021916.3 2917 UTR 3 NP_068735.1

View Full Product Details