Product Details

SNP ID
rs12188417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140719011 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGACCCGCGGGTGCTTTCCAGCTCT[G/T]TTACTGCTGAAGTTCAGCTCCTTTT
Phenotype
MIM: 612695 MIM: 612696 MIM: 612697
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
VTRNA1-1 PubMed Links

Gene Details

Gene
VTRNA1-1
Gene Name
vault RNA 1-1
There are no transcripts associated with this gene.

Gene
VTRNA1-2
Gene Name
vault RNA 1-2
There are no transcripts associated with this gene.

Gene
VTRNA1-3
Gene Name
vault RNA 1-3
There are no transcripts associated with this gene.

View Full Product Details