Product Details

SNP ID
rs12109317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140704845 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTTTTTTTTGGTCATACAGATA[C/T]TTCTCAACTTCTGATGGGGTTACCT
Phenotype
MIM: 600783 MIM: 612695
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HARS2 PubMed Links

Gene Details

Gene
HARS2
Gene Name
histidyl-tRNA synthetase 2, mitochondrial
There are no transcripts associated with this gene.

Gene
VTRNA1-1
Gene Name
vault RNA 1-1
There are no transcripts associated with this gene.

Gene
ZMAT2
Gene Name
zinc finger matrin-type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144723.2 Intron NP_653324.1

View Full Product Details