Product Details

SNP ID
rs12148878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:83656656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATCTAAAGGCAGGGCACATCTTA[A/G]TATTGTGTCCCAAGGGCTGCTGCTA
Phenotype
MIM: 609199
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAMTSL3 PubMed Links

Gene Details

Gene
ADAMTSL3
Gene Name
ADAMTS like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301110.1 Intron NP_001288039.1
NM_207517.2 Intron NP_997400.2
XM_011521822.2 Intron XP_011520124.1
XM_011521823.2 Intron XP_011520125.1
XM_011521824.2 Intron XP_011520126.1
XM_011521825.2 Intron XP_011520127.1
XM_017022434.1 Intron XP_016877923.1
XM_017022435.1 Intron XP_016877924.1

View Full Product Details