Product Details

SNP ID
rs11141322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:86267437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGTTTAATAAGAAAGCACACTTA[A/G]GAATCTAGGCCTCATTCTCCCATGT
Phenotype
MIM: 611006
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf153 PubMed Links

Gene Details

Gene
C9orf153
Gene Name
chromosome 9 open reading frame 153
There are no transcripts associated with this gene.

Gene
ISCA1
Gene Name
iron-sulfur cluster assembly 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030940.3 Intron NP_112202.2

View Full Product Details