Product Details

SNP ID
rs10465156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92158213 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGTTTGAAGAGCCCTAATCAGGT[A/G]CATTGCAGGACGCTCCTCAGTTGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00475 PubMed Links

Gene Details

Gene
LINC00475
Gene Name
long intergenic non-protein coding RNA 475
There are no transcripts associated with this gene.

View Full Product Details