Product Details

SNP ID
rs12457901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:62734185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATGAGGTTGATGGGAAGTGCTG[C/T]TTATCTTCTGAAGGGAGGTACTTTT
Phenotype
MIM: 609396
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHLPP1 PubMed Links

Gene Details

Gene
PHLPP1
Gene Name
PH domain and leucine rich repeat protein phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194449.3 Intron NP_919431.2
XM_011525886.1 Intron XP_011524188.1

View Full Product Details