Product Details

SNP ID
rs11995570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:39921211 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGATTAATTTTTTTTTTTAAAGAT[C/T]GCTGGGAAAAGTGGGTGAATCACCT
Phenotype
MIM: 147435
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IDO1 PubMed Links

Gene Details

Gene
IDO1
Gene Name
indoleamine 2,3-dioxygenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002164.5 Intron NP_002155.1

View Full Product Details